Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gephyrin/GPHN Mouse anti-Human, Monkey, Allophycocyanin/Cy7, Clone: OTI3B6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271556APCCY7

Catalog No. N27156APCY7


Add to cart

Description

Description

Gephyrin/GPHN Monoclonal antibody specifically detects Gephyrin/GPHN in Human, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

Gephyrin/GPHN
OTI3B6
Flow Cytometry
Affinity Purified, Conjugated
GPHN
Full length recombinant protein of human Gephyrin/GPHN (NP_065857) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Monkey
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
gephyrin, GPHRYN, KIAA1385GPHGEPH
Mouse
IgG2a
0.1 mL
Primary
10243.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit