Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gephyrin/GPHN Rabbit anti-Human, Mouse, Rat, Clone: JG34-47, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275504

Catalog No. NBP275504


Add to cart

Description

Description

Gephyrin/GPHN Monoclonal antibody specifically detects Gephyrin/GPHN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Gephyrin/GPHN
JG34-47
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
gephyrin, GPHRYN, KIAA1385GPHGEPH
Rabbit
IgG
Protein A purified
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin
Purified
GPHN
Recombinant protein within human Gephyrin aa 1-100.
80 kDa
100 μL
Primary
10243.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit