Geranyl acetate, 98%

Manufacturer: Alfa Aesar A1986422

Quantity 100g

 View more versions of this product

Catalog No. AAA1986422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 105-87-3
    Molecular Formula (CH3)2=CHCH2CH2C(CH3)=CHCH2O2CCH3
    Formula Weight 196.29
    MDL Number MFCD00015037
    Chemical Name or Material Geranyl acetate

    Specifications

    Formula Weight 196.29
    Quantity 100g
    TSCA TSCA
    MDL Number MFCD00015037
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present
    Recommended Storage 2° to 8°C
    RTECS Number RG5920000
    Density 0.91
    Molecular Formula (CH3)2=CHCH2CH2C(CH3)=CHCH2O2CCH3
    CAS 105-87-3
    Assay Percent Range 0.98
    Chemical Name or Material Geranyl acetate
    Assay 98%

    Description

    Applications
    It is used primarily as a component of perfumes for creams and soaps and as a flavoring ingredient. It is used particularly in rose, lavender and geranium formulations where a sweet fruity or citrus aroma is desired. It is found to have has anti-oxidant, anti-inflammatory and anti-proliferative activities.

    Solubility
    Not miscible or difficult to mix in water.

    Notes
    Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    2° to 8°C

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit