Learn More
Geranyl bromide, 96%, Thermo Scientific Chemicals
Description
Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.
Solubility
Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Density | 1.094 |
Boiling Point | 99°C to 101°C (10 mmHg) |
Flash Point | 95°C (203°F) |
Refractive Index | 1.504 |
Quantity | 5 g |
Beilstein | 1422412 |
Sensitivity | Light sensitive |
Solubility Information | Immiscible with water. |
Formula Weight | 217.16 |
Percent Purity | 96% |
Show More |
Safety and Handling
Hazard Category | H227-H315-H318-H335 |
Hazard Statement | GHS H Statement H318-H335-H227-H315 Causes serious eye damage. May cause respiratory irritation. Combustible liquid. Causes skin irritation. |
Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c |
EINECSNumber | 228-123-7 |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.