Learn More
Germaine™ Laboratories Febrile Antigens For Salmonella H
Febrile antigens intended for detection of bacterial agglutinins associated with Salmonella H infection
Manufacturer:  GERMAINE LABORATORIES 1402
 View more versions of this product
Catalog No. 16-414-119
  Check Availability
Add to cart
Disclaimers:
Professional use only
Specifications Description & Specifications
Specifications
Salmonella H C Antibodies
5mL
Salmonella H c Antigen