Germaine™ Labs AimStrip™ Hb Hemoglobin (Hb) Test System

Provides lab quality accuracy for hemoglobin and hematocrit testing

Manufacturer: GERMAINE LABORATORIES 78150

 View more versions of this product

Catalog No. 23-111-283

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Sample Type Capillary and venous whole blood
    Includes 50 Test cartridges, 1 code chip
    Sample Size 10μL
    Type Hemoglobin Test Pack
    For Use With (Application) Hb and Hct testing
    Battery Life 2700 tests or 360 hours
    Calibration Type Automatic and factory calibrated
    Memory 1000 Tests, date/time and ID number
    Power Supply Details 4 AAA batteries or AC adaptor
    Range Hb: 5.6 to 23.5g/dL
    Regulatory Status CLIA waived
    Time Range <15 seconds

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit