Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germanium, 99.999%, (trace metal basis), powder, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Glass bottle
7440-56-4
Ge
72.59
MFCD00085310
GNPVGFCGXDBREM-UHFFFAOYSA-N
element,germanio,germanium, metal powder,unii-00072j7xws,powder,germanium, chips trace metals basis,atom,copper metal,germanium, disks, 15mm, thickness 1.0mm, polycrystalline,germanium, powder
6326954
CHEBI:30441
germanium
[Ge]
7440-56-4
Ge
72.59
MFCD00085310
GNPVGFCGXDBREM-UHFFFAOYSA-N
element,germanio,germanium, metal powder,unii-00072j7xws,powder,germanium, chips trace metals basis,atom,copper metal,germanium, disks, 15mm, thickness 1.0mm, polycrystalline,germanium, powder
6326954
CHEBI:30441
germanium
[Ge]
For ultra trace analysis
Assay Percent Range 99.999%
Merck Index 15, 4441
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in hcl and diluted alkali hydroxides
Formula Weight 72.59
Physical Form Glistening Crystalline Powder
Percent Purity 99.999%
Grade Trace Metal Basis
Density 5.3500g/mL
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: insoluble in hcl and diluted alkali hydroxides
Specific Gravity 5.35
Total Trace Metal Impurities 0.001% max.
Boiling Point 2830.0°C
Color Black to Gray
Melting Point 937.0°C
Quantity 50g
Chemical Name or Material Germanium, 99.999%
Show More Show Less

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-164-3

RTECSNumber : LY5200000

TSCA : TSCA