Alfa Aesar™ Germanium(IV) chloride, 99.9999% (metals basis)

Manufacturer: Alfa Aesar™ 1050902

Quantity 0.5g

 View more versions of this product

Catalog No. AA1050902

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10038-98-9
    Linear Formula GeCl4
    Formula Weight 214.4
    MDL Number MFCD00011029
    UN Number UN3264
    Merck Index 14,4410
    Chemical Name or Material Germanium(IV) chloride

    Specifications

    Formula Weight 214.4
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011029
    Melting Point −49.5°C
    Quantity 0.5g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Density 1.874g/mL
    Physical Form Liquid
    UN Number UN3264
    Boiling Point 83.1°C
    Odor Acidic
    Sensitivity Moisture Sensitive
    Merck Index 14,4410
    Color Colorless
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
    TSCA TSCA
    Linear Formula GeCl4
    CAS 10038-98-9
    RTECS Number LY5225000
    Chemical Name or Material Germanium(IV) chloride
    Assay 99.9999%, (metals basis)

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit