Alfa Aesar™ Germanium(IV) ethoxide, 97%

Manufacturer: Alfa Aesar™ 1467914

Quantity 25g

 View more versions of this product

Catalog No. AA1467914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14165-55-0
    Linear Formula Ge(OC2H5)4
    Formula Weight 252.84
    MDL Number MFCD00015121
    UN Number UN1993
    Synonym Germanium tetraethoxide; Tetraethoxygermane
    Chemical Name or Material Germanium(IV) ethoxide

    Specifications

    Formula Weight 252.84
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338-P337+P313
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Physical Form Liquid
    Color Colorless
    Odor Alcohol-like
    Sensitivity Moisture Sensitive
    TSCA TSCA
    CAS 14165-55-0
    Quantity 25g
    UN Number UN1993
    Synonym Germanium tetraethoxide; Tetraethoxygermane
    Solubility Information Soluble in anhydrous alcohol and benzene; Hydrolyzes in water
    Flash Point 49° (121°F)
    Health Hazard 1 Warning
    Density 1.14g/mL
    Boiling Point 185.5°C
    Melting Point −72°C
    Linear Formula Ge(OC2H5)4
    MDL Number MFCD00015121
    Health Hazard 2 GHS H Statement
    H226-H315-H319
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    Refractive Index 1.4070
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material Germanium(IV) ethoxide
    Assay 97%

    GHS H Statement
    H226-H315-H319
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338-P337+P313
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit