Alfa Aesar™ Germanium(IV) isopropoxide, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 4267314

Quantity 25g

 View more versions of this product

Catalog No. AA4267314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21154-48-3
    Linear Formula Ge[OCH(CH3)2]4
    Formula Weight 308.94
    MDL Number MFCD00015339
    Synonym Tetraisopropoxygermane
    Chemical Name or Material Germanium (IV) isopropoxide

    Specifications

    Formula Weight 308.94
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338-P337+P313
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Boiling Point 109°C (30mmHg)
    Physical Form Liquid
    Quantity 25g
    Density 1.025g/mL
    Odor Alcohol-like
    Sensitivity Moisture Sensitive
    MDL Number MFCD00015339
    CAS 21154-48-3
    Synonym Tetraisopropoxygermane
    Health Hazard 1 Warning
    Refractive Index 1.414
    Linear Formula Ge[OCH(CH3)2]4
    Flash Point 68°C (154°F)
    Health Hazard 2 GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.
    Color Colorless
    Chemical Name or Material Germanium (IV) isopropoxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.

    GHS P Statement
    P280g-P305+P351+P338-P337+P313
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit