Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Germanium Optical Window, 50.8mm (2.0 in.) dia x 1mm (0.04 in.) thick

Manufacturer:  Thermo Scientific™ 45388KS

Catalog No. AA45388KS


Order in Bulk

Request bulk or custom formats

Add to cart

Description

Description

Specifications

Specifications

Germanium Optical Window
7440-56-4
MFCD00085310
GNPVGFCGXDBREM-UHFFFAOYSA-N
germanium
6326954
CHEBI:30441
1 ea.
Ge
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
[Ge]
72.63
Shrink-wrapped
Uncoated,Clear Aperture 85%,Surface Finish 40/20
Safety and Handling

Safety and Handling

EINECSNumber : 231-164-3

TSCA : Yes

SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit