Germanium powder, 99.99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1019009

Quantity 10g

 View more versions of this product

Catalog No. AA1019009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-56-4
    Physical Form Powder
    Assay Percent Range 99.9999% (Metals basis)
    MDL Number MFCD00085310
    Chemical Name or Material Germanium

    Specifications

    Quantity 10g
    Color Silver
    Density 5.32g/cm3
    EINECS Number 231-164-3
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.9999% (Metals basis)
    Appearance Silver-colored powder
    MDL Number MFCD00085310
    CAS 7440-56-4
    Vapor Pressure 0 hPa
    Chemical Name or Material Germanium

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit