Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germanium tetrachloride, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
10038-98-9
Cl4Ge
214.4
MFCD00011029
IEXRMSFAVATTJX-UHFFFAOYSA-N
germanium tetrachloride,germanium chloride,germane, tetrachloro,germanium iv chloride,germanium, tetrachloro,gecl4,germanium chloride gecl4,unii-ysv1r803c0,wln: .ge..g4,germanium chloride gecl4 8ci
66226
tetrachlorogermane
Cl[Ge](Cl)(Cl)Cl
10038-98-9
Cl4Ge
214.4
MFCD00011029
IEXRMSFAVATTJX-UHFFFAOYSA-N
germanium tetrachloride,germanium chloride,germane, tetrachloro,germanium iv chloride,germanium, tetrachloro,gecl4,germanium chloride gecl4,unii-ysv1r803c0,wln: .ge..g4,germanium chloride gecl4 8ci
66226
tetrachlorogermane
Cl[Ge](Cl)(Cl)Cl
for ultra trace analysis
Assay Percent Range 99.99%
Linear Formula GeCl4
Merck Index 15, 4444
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in benzene,ether,carbon disulfid and,other organic solvents,insoluble in concentrated hydrochloric acid
Formula Weight 214.4
Physical Form Liquid
Percent Purity 99.99%
Grade Trace Metal Basis
Density 1.8400g/mL
Packaging Glass bottle
Solubility Solubility in water: decomposes. Other solubilities: soluble in benzene, ether, carbon disulfid and, other organic solvents, insoluble in concentrated hydrochloric acid
Specific Gravity 1.84
Total Trace Metal Impurities 0.01% max.
Viscosity 529 mPa.s (37°C)
Boiling Point 82.0°C to 84.0°C
Color Colorless
Melting Point -49.5°C
Quantity 5g
Chemical Name or Material Germanium tetrachloride, 99.99%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
May cause respiratory irritation.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 233-116-7

RTECSNumber : LY5225000

TSCA : TSCA