Learn More
Getinge USA Getinge Clean Tec Rinse, 4x1gal  DFS Item
Manufacturer:  Getinge USA 61301605280
Scale fighting rinse additive designed to be injected into the final freshwater rinse of automatic washers. Reduces hard water spotting and reduces drying time for carts, cases and instruments.
Catalog No. NC1376453