Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ GFAP (Glial Fibrillary Acidic Protein), Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ GFAP (Glial Fibrillary Acidic Protein), Rabbit Polyclonal Antibody.

$215.50 - $809.50

Specifications

Antigen GFAP (Glial Fibrillary Acidic Protein)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
RB087A
View Documents
Thermo Scientific™
RB087A
1mL
Each for $809.50
Add to cart
 
RB087A0
View Documents
Thermo Scientific™
RB087A0
100μL
Each for $215.50
Add to cart
 
RB087A1
View Documents
Thermo Scientific™
RB087A1
500μL
Each for $568.00
Add to cart
 
RB087R7
View Documents
Thermo Scientific™
RB087R7
7mL
Each for $354.00
Add to cart
 
RB087R1
View Documents
Thermo Scientific™
RB087R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ GFAP (Glial Fibrillary Acidic Protein), Rabbit Polyclonal Antibody.

Glial Fibrillary Acidic Protein (GFAP) is specific to astrocytes (i.e., glial cells) and ependymal cells of the central nervous system.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey, Cow, Sheep, Guinea Pig, Chicken, Mouse, Rat and Hamster. Others not known.

Epitope: Not determined

Immunogen: GFAP isolated from cow spinal cord

Molecular Weight: 51-52 kDa

Positive Control: Brain or astrocytoma

Cellular Localization: Cytoplasmic Ab-4 stains astrocytes, glial cells, ependymal cells and their corresponding tumors. In the peripheral nervous system, Schwann cells, satellite cells, and enteric glial cells are stained. Weak staining of axons has been observed. Absorption studies show that this is caused by a cross-reaction with neurofilament protein. Ab-4 is particularly useful for distinguishing neoplasms of astrocytic origin from other neoplasms in the central nervous system by immunohistochemistry.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

GFAP (Glial Fibrillary Acidic Protein)
Polyclonal
Rabbit
Neuroscience
GFAP isolated from cow spinal cord
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Primary
IgG
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit