Learn More
GFAP Mouse anti-Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit, Alexa Fluor™ 350, Clone: GA5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233184AF350
Catalog No. NP233184A30
Add to cart
Description
Description
GFAP Monoclonal antibody specifically detects GFAP in Human,Mouse,Rat,Porcine,Bovine,Chicken,Rabbit samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
GFAP
GA5
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
FLJ45472, GFAP astrocytes, glial fibrillary acidic protein
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
This MAb recognizes a protein of ~50kDa which is identified as Glial Fibrillary Acidic Protein (GFAP). It shows no cross-reaction with other intermediate filament proteins. GFAP is specifically found in astroglia. GFAP is a very popular marker for localizing benign astrocyte and neoplastic cells of glial origin in the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
GFAP
Glial fibrillary protein from porcine spinal cord was used as the immunogen for the GFAP (GA5) antibody.
0.1 mL
Primary
2670
Bovine, Chicken, Human, Mouse, Porcine, Rabbit, Rat