Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP, Mouse anti-Pig, Clone: GFA-02, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB16155

Catalog No. 89-998-929

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

GFAP
GFA-02
Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Immunohistochemistry (Frozen sections) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
glial fibrillary acidic protein
-
Mouse
IgG1
RUO
Antibody
Monoclonal
Human, Pig
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
In PBS (0.05% sodium azide).
Liquid
GFAP
Crude preparation from porcine spinal cord.
50 μg
Store at 4°C.
Primary
396562
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit