Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP Rabbit anti-Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit, Clone: ASTRO/1974R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30787710UG

 View more versions of this product

Catalog No. NB123227


Add to cart

Description

Description

GFAP Monoclonal specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin.
Specifications

Specifications

GFAP
ASTRO/1974R
Unconjugated
10 mM PBS with 0.05% BSA
ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein
This GFAP Antibody (ASTRO/1974R) was developed against recombinant human full-length GFAP protein.
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit
Flow Cytometry, Immunohistochemistry (Paraffin)
0.2 mg/ml
Flow Cytometry 1 - 2 ug/million cells, Immunohistochemistry-Paraffin 1 - 2 ug/mL
Purified
Rabbit
IgG
100 μg
Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
Primary
2670
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit