Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ GFP (Green Fluorescent Protein) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results with Thermo Scientific™ GFP (Green Fluorescent Protein) Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS1288P0

 View more versions of this product

Catalog No. MS1288P0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ensure accurate, reproducible results with Thermo Scientific™ GFP (Green Fluorescent Protein) Ab-1, Mouse Monoclonal Antibody.

GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species-independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expression and protein localization. Moreover, other applications for GFP include its use in assessing protein-protein interactions in the yeast two-hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.

Host Species: Mouse

Clone: GFP01; same as B34

Isotype: IgG1/κ

Species Reactivity: Proteins from all species that are tagged with GFP.

Epitope: Not determined

Immunogen: Purified recombinant GFP protein

Molecular Weight: 27 kDa

Positive Control: Purified recombinant GFP protein, transfected cell lines expressing GFP-tagged proteins

Cellular Localization: Depends upon the localization of the parent protein tagged with GFP

Recommended for:

  • Western Blotting (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

GFP (Green Fluorescent Protein) Ab-1
GFP01 (B34)
Unconjugated
Murine
IgG2a kappa
RUO
Primary
Monoclonal
Monoclonal
200μg/mL
Concentrated
Purified recombinant GFP protein
100μL
Epitope Tags
Primary
All
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit