Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Mouse anti-Non-species specific, FITC, Clone: 4B10B2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222111F
Description
GFP Monoclonal specifically detects GFP in Non-species specific samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Product Image.Specifications
GFP | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Nonspecies specific | |
Purified |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
4B10B2 | |
Flow Cytometry, ELISA FLOW CYTOMETRYIMMUNOCYTOCHEMISTRY/IMMUNOFLUORESCENCE, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
Purified recombinant fragment of GFP expressed in E. coli. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only