Learn More
GFP Mouse, BV421, Clone: 1A12-6-18, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566040
Catalog No. BDB566040
Add to cart
Description
Description
GFP Monoclonal antibody specifically detects GFP in samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
Specifications
GFP
1A12-6-18
Brilliant Violet 421
Purified
Mouse IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV421 were removed.
Monoclonal
Flow Cytometry, Flow Cytometry (Intracellular Staining)
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
50μg
Cell Biology
Primary
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.