Learn More
GFR alpha-3/GDNF R alpha-3 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0310-2P25, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212337AF532
Catalog No. NP212337A32
Add to cart
Description
Description
GFR alpha-3/GDNF R alpha-3 Monoclonal antibody specifically detects GFR alpha-3/GDNF R alpha-3 in Human samples. It is validated for Western Blot.
Specifications
Specifications
GFR alpha-3/GDNF R alpha-3
MM0310-2P25
Western Blot
GDNF family receptor alpha 3, GDNF family receptor alpha-3, GDNF receptor alpha-3, GDNFR-alpha-3, GFRa-3, GFR-alpha-3, glial cell line-derived neurotrophic factor receptor alpha-3, GPI-linked receptor
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
GFRA3
Human GFR alpha 3
0.1 mL
Primary
2676