Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFR alpha-3/GDNF R alpha-3 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0310-2P25, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212337AF594

Catalog No. NP212337A94


Add to cart

Description

Description

GFR alpha-3/GDNF R alpha-3 Monoclonal antibody specifically detects GFR alpha-3/GDNF R alpha-3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

GFR alpha-3/GDNF R alpha-3
MM0310-2P25
Western Blot
Conjugated, Purified
GFRA3
Human GFR alpha 3
Protein G purified
Cytokine Research, Immunology
Primary
2676.0
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
GDNF family receptor alpha 3, GDNF family receptor alpha-3, GDNF receptor alpha-3, GDNFR-alpha-3, GFRa-3, GFR-alpha-3, glial cell line-derived neurotrophic factor receptor alpha-3, GPI-linked receptor
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit