Learn More
GFR alpha-4/GDNF R alpha-4/Persephin receptor Rat anti-Mouse, Alexa Fluor 700, Clone: RM0244-1M21, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212415AF700
Catalog No. NB212415700
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
GFR alpha-4/GDNF R alpha-4/Persephin receptor Monoclonal antibody specifically detects GFR alpha-4/GDNF R alpha-4/Persephin receptor in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
GFR alpha-4/GDNF R alpha-4/Persephin receptor
RM0244-1M21
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
AAI41875.1 GI
GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR receptor alpha 4, GFR-alpha-4, Persephin receptor
Rat
IgG2
0.1 ml
Primary
64096
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Purified
GFRA4
Mouse
Protein G purified
Store at 4C in the dark.
Monoclonal
Mouse