Learn More
GFR alpha-4/GDNF R alpha-4/Persephin receptor Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0244-1M21, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212415AF750
Catalog No. NP21241A70
Add to cart
Description
Description
GFR alpha-4/GDNF R alpha-4/Persephin receptor Monoclonal antibody specifically detects GFR alpha-4/GDNF R alpha-4/Persephin receptor in Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
GFR alpha-4/GDNF R alpha-4/Persephin receptor
RM0244-1M21
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR receptor alpha 4, GFR-alpha-4, Persephin receptor
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
GFRA4
Mouse
0.1 mL
Primary
64096