Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFR alpha-like Rabbit anti-Human, Alexa Fluor™ 350, Clone: 2345C, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB9697U100UG
This item is not returnable.
View return policy
Description
GFR alpha-like Monoclonal specifically detects GFR alpha-like in Human samples. It is validated for Flow Cytometry.Specifications
GFR alpha-like | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Human | |
Supernatant |
Flow Cytometry | |
2345C | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q6UXV0 | |
Human embryonic kidney cell line HEK293-derived recombinant human GFR alpha-like, Ser19-Glu351, Accession # Q6UXV0 | |
100 μg | |
Detects human GFR alpha-like in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title