Learn More
GFR alpha-like Rabbit anti-Human, Alexa Fluor™ 750, Clone: 2345C, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB9697S100UG
Catalog No. FAB9697S100
Add to cart
This item is not returnable. View return policy
Description
Description
GFR alpha-like Monoclonal antibody specifically detects GFR alpha-like in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
GFR alpha-like
2345C
Flow Cytometry 0.25-1 μg/106 cells
Q6UXV0
Rabbit
IgG
100 μg
Primary
Detects human GFR alpha-like in direct ELISAs.
Flow Cytometry
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Human embryonic kidney cell line HEK293-derived recombinant human GFR alpha-like, Ser19-Glu351, Accession # Q6UXV0
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Human