Learn More
GFR alpha-like Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP30960625UL
 View more versions of this product
Catalog No. NB124114
Add to cart
Description
Description
GFR alpha-like Polyclonal antibody specifically detects GFR alpha-like in Human samples. It is validated for Western Blot
Specifications
Specifications
GFR alpha-like
Unconjugated
PBS buffer, 2% sucrose
Rabbit
Affinity purified
RUO
Primary
Human
Western Blot
Western Blot 1.0 ug/ml
Purified
The immunogen is a synthetic peptide directed towards the N-terminal region of Human GFR alpha-like (NP_997293). Peptide sequence TDDFYCTVNKLLGKKCINKSDNVKEDKFKWNLTTRSHHGFKGMWSCLEVA
25 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal