Sino Biological™ GFRA1 Recombinant Rat Protein, His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding rat GFRA1 (Met 1-Leu 445) fused to polyhistidine tag at C-terminus

Manufacturer: Sino Biological™ 80021R08H25

 View more versions of this product

Catalog No. 80-021-R08H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 48kd
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Source Rat
    Structural Form Full Length, Recombinant
    Endotoxin Concentration <1 EU/μg
    Common Name GDNF Family Receptor Alpha 1
    Gene Symbol GFRA1
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Growth Factors and Receptors
    Expression System Mammalian
    Product Type GFRA1 Recombinant Rat Protein
    For Use With (Application) Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 25454

    Expressed from DNA sequence encoding rat GFRA1 (Met 1-Leu 445) fused to polyhistidine tag at C-terminus

    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit