GFS Chemicals Amco Clear™ Linear Calibration Sets

Highly accurate, stable and reproducible standards for calibrating turbidimeters and nephelometers.

Manufacturer: GFS Chemicals 85566

 View more versions of this product

Catalog No. 15-426-10

  Check Availability
Add to Cart

    Specifications

    Turbidity 0, 0.2, 0.4, 0.6, 0.8, 1 NTU
    Includes 125mL of each 0, 0.2, 0.4, 0.6, 0.8 and 1.0 NTU primary standards
    Type Linear Calibration Kit
    Certifications/Compliance EPA
    Shelf Life 1 Year
    Accuracy 1%

    • Ready to use — no mixing or dilution required
    • Consist of polymer microspheres suspended in a pure, aqueous medium
    • Uniform in particle size distribution to ensure consistent light scattering and precise enough to permit calibration down to and including 0 NTU
    • Nontoxic and won't settle out of suspension
    • Guaranteed accuracy within 1%
    • Designed for 90° white light turbidimeters (EPA 180.1 design)
    • Guaranteed one-year shelf life

    Order Info

    Available in three ranges. Each set consists of six 125mL bottles of primary standards.

    Compliance

    Fulfill U.S. EPA recommendations for linear calibration of each range used to permit instrument-to-instrument correlation

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit