Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GGA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102379
Description
Antibody detects endogenous levels of total GGA2.
This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding (GGA) family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. This family member may play a significant role in cargo molecules regulation and clathrin-coated vesicle assembly.
Specifications
GGA2 | |
Polyclonal | |
Unconjugated | |
GGA2 | |
1200007E24Rik; ADP-ribosylation factor binding protein 2; ADP-ribosylation factor-binding protein GGA2; gam; Gamma-adaptin-related protein 2; Gga2; golgi associated, gamma adaptin ear containing, ARF binding protein 2; golgi-associated, gamma adaptin ear containing, ARF binding protein 2; golgi-localized, gamma ear-containing, ARF-binding protein 2; Kiaa1080; mKIAA1080; VEAR; VHS domain and ear domain of gamma-adaptin; VHS domain and ear domain-containing protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
23062, 293455, 74105 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6P5E6, Q9UJY4 | |
GGA2 | |
A synthesized peptide derived from human GGA2(Accession Q9UJY4), corresponding to amino acid residues V529-P579. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction