Learn More
GGA2Rabbit anti-Human, Polyclonal, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 600401849

Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use.
This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding (GGA) family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. This family member may play a significant role in cargo molecules regulation and clathrin-coated vesicle assembly.Specifications
GGA2 | |
Polyclonal | |
Unconjugated | |
GGA2 | |
1200007E24Rik; ADP-ribosylation factor binding protein 2; ADP-ribosylation factor-binding protein GGA2; gam; Gamma-adaptin-related protein 2; Gga2; golgi associated, gamma adaptin ear containing, ARF binding protein 2; golgi-associated, gamma adaptin ear containing, ARF binding protein 2; Golgi-localized, gamma ear-containing, ARF-binding protein 2; KIAA1080; mKIAA1080; Vear; VHS domain and ear domain of gamma-adaptin; VHS domain and ear domain-containing protein | |
Rabbit | |
RUO | |
23062 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1.4 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
Q9UJY4 | |
GGA2 | |
100 μg | |
Primary | |
Human | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.