Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGN Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549325

Catalog No. PA549325


Add to cart

Description

Description

May be involved in spermatogenesis.
Specifications

Specifications

GGN
Polyclonal
PBS with 0.09% sodium azide
Q86UU5
Gametogenetin, GGN
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
199720
Western Blot
Unconjugated
GGN
Liquid
GGN
KLH conjugated synthetic peptide between 139-163 amino acids from the Central region of human GGN.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit