Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GGN Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | GGN |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP153063
|
Novus Biologicals
NBP153063 |
100 μL |
Each for $436.00
|
|
NBP15306320
|
Novus Biologicals
NBP15306320UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
GGN Polyclonal specifically detects GGN in Human samples. It is validated for Western Blot.Specifications
GGN | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ35713, gametogenetin, MGC33369 | |
GGN | |
IgG | |
Affinity Purified | |
67 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q86UU5 | |
199720 | |
Synthetic peptides corresponding to GGN(gametogenetin) The peptide sequence was selected from the N terminal of GGN. Peptide sequence GPSKWQKPAGTPVPRIRRLLEASHRGQGDPPSLRPLKPPPPPRQLSVKDT. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title