Learn More
GGPS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 149441AP
Catalog No. 50-172-9853
Add to cart
Description
Description
This gene is a member of the prenyltransferase family and encodes a protein with geranylgeranyl diphosphate (GGPP) synthase activity. The enzyme catalyzes the synthesis of GGPP from farnesyl diphosphate and isopentenyl diphosphate. GGPP is an important molecule responsible for the C20-prenylation of proteins and for the regulation of a nuclear hormone receptor. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
GGPS1
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95749, Q6F596, Q9WTN0
GGPP synthase, GGPPS, GGPPS1, GGPPSase, GGPS1
Rabbit
IgG
150 μL
-20°C
Primary
14593, 291211, 9453
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
GGPS1
Liquid
Ggps1
GGPS1 Fusion Protein Ag6745
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat