Learn More
Thermo Scientific™ GGT Reagents
Determine quantity of gamma-glutamyl transferase (GGT) in human serum or plasma with these reagents.
$333.00
Specifications
CE Marker Yes
Methods Compatibility Kinetic
Storage Requirements 2°C to 8°C
View More Specs
Products 1
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
TR19110
Accessories View Documents
20 x 10mL
Pack of 20 for $333.00
Add to cart
Description
Description
Determine quantity of gamma-glutamyl transferase (GGT) in human serum or plasma in vitro with Thermo Scientific™ GGT Reagents.
GGT present in the serum originates primarily from the hepatobiliary system, and its activity is elevated in all forms of liver disease. Increased concentrations of the enzyme are also found in chronic alcoholism and with anticonvulsant therapy.
  • Reagents offer the laboratory 21 days reconstituted stability
Specifications
Specifications
Yes
Kinetic
2°C to 8°C