Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGT2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548465

Catalog No. PA548465


Add to cart

Description

Description

Isoform 1, isoform 2 and isoform 3 lack catalytic activity due to its inability to undergo the autocatalytic cleavage needed to produce a mature, enzymatically active heterodimer.
Specifications

Specifications

GGT2
Polyclonal
PBS with 0.09% sodium azide
P36268
Inactive gamma-glutamyltranspeptidase 2, GGT 2, Gamma-glutamyltransferase 2, Glutathione hydrolase 2, GGT2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
728441
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GGT2
Liquid
GGT2
KLH conjugated synthetic peptide between 12-40 amino acids from the N-terminal region of human GGT2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit