Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GGT7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543301
Description
Peptide sequence: ILLNSQMLDF SWPNRTANHS APSLENSVQP GKRPLSFLLP TVVRPAEGLC Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%.
GGTL3 is an enzymes involved in both the metabolism of glutathione and in the transpeptidation of amino acids. Changes in the activity of gamma-glutamyltransferase may signal preneoplastic or toxic conditions in the liver or kidney. GGTL3 consists of a heavy and a light chain, and it can interact with CT120, a plasma membrane-associated protein that is possibly involved in lung carcinogenesis.This gene is a member of a gene family that encodes enzymes involved in both the metabolism of glutathione and in the transpeptidation of amino acids. Changes in the activity of gamma-glutamyltransferase may signal preneoplastic or toxic conditions in the liver or kidney. The protein encoded by this gene consists of a heavy and a light chain, and it can interact with CT120, a plasma membrane-associated protein that is possibly involved in lung carcinogenesis.
Specifications
GGT7 | |
Polyclonal | |
Unconjugated | |
Ggt7 | |
1110017C11Rik; 6330563L03Rik; D20S101; dJ18C9.2; gamma-glutamyltransferase 4; gamma-glutamyltransferase 7; gamma-glutamyltransferase 7; glutathione hydrolase 7; gamma-glutamyltransferase-like 3; Gamma-glutamyltransferase-like 5; gamma-glutamyltranspeptidase 7; gamma-glutamyltranspeptidase-like 3; GGT 7; GGT4; Ggt7; GGTL3; GGTL5; Glutathione hydrolase 7; Glutathione hydrolase 7 heavy chain; Glutathione hydrolase 7 light chain; LOC100732094; mFLJ00311 protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2686 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UJ14 | |
Ggt7 | |
Synthetic peptide directed towards the C-terminal of human GGTL3 (aa 506-555). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction