Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GGT7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564947
Description
Immunogen sequence: HLVLSPPPPHT GPALISALNI LEGFNLTSLV SREQALHWVA ETLKIALALA SRLGDPVYDS TITESMDDML SKVEAAYLRG HINDSQAAPA PLLPVYELDG APTAAQVLIM GPDDFIVAMV SSLNQPFGSG LITPSGILLN S Highest antigen sequence identity to the following orthologs - mouse 96%, rat 95%.
GGTL3 is an enzymes involved in both the metabolism of glutathione and in the transpeptidation of amino acids. Changes in the activity of gamma-glutamyltransferase may signal preneoplastic or toxic conditions in the liver or kidney. GGTL3 consists of a heavy and a light chain, and it can interact with CT120, a plasma membrane-associated protein that is possibly involved in lung carcinogenesis.This gene is a member of a gene family that encodes enzymes involved in both the metabolism of glutathione and in the transpeptidation of amino acids. Changes in the activity of gamma-glutamyltransferase may signal preneoplastic or toxic conditions in the liver or kidney. The protein encoded by this gene consists of a heavy and a light chain, and it can interact with CT120, a plasma membrane-associated protein that is possibly involved in lung carcinogenesis.
Specifications
GGT7 | |
Polyclonal | |
Unconjugated | |
Ggt7 | |
1110017C11Rik; 6330563L03Rik; D20S101; dJ18C9.2; gamma-glutamyltransferase 4; gamma-glutamyltransferase 7; gamma-glutamyltransferase 7; glutathione hydrolase 7; gamma-glutamyltransferase-like 3; Gamma-glutamyltransferase-like 5; gamma-glutamyltranspeptidase 7; gamma-glutamyltranspeptidase-like 3; GGT 7; GGT4; Ggt7; GGTL3; GGTL5; Glutathione hydrolase 7; Glutathione hydrolase 7 heavy chain; Glutathione hydrolase 7 light chain; LOC100732094; mFLJ00311 protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2686 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UJ14 | |
Ggt7 | |
Recombinant Human GGT7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction