Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GH1 Mouse anti-Human, Clone: SPM106, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14800

Catalog No. 89-034-883


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human GH1.

  • High specificity
  • High reactivity
Specifications

Specifications

GH1
SPM106
Mouse monoclonal antibody raised against partial recombinant human GH1.
In 10 mM PBS.
Liquid
GH1
Recombinant protein corresponding to amino acids 58-187 of human GH1.
Protein A/G purification
RUO
Primary
2688
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
P01241
GH|GH-N|GHN|hGH-N
Mouse
IgG2b
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)