Learn More
Andwin ScientificSupplier Diversity Partner GHP .45MICRON 47MM DISC 100/PK DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner 66548
The GH Polypro (GHP) Membrane Disc Filters feature maximum chemical compatibility when filtering both aqueous solutions and aggressive solvents. The low protein binding of this product offers high recovery of critical proteinaceous samples. The filters have pore size of 0.45 �m and have diameter of 47 mm. It is sold in a pack of 100.
Catalog No. NC0134003