Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ghrelin Receptor/GHSR Mouse anti-Human, Alexa Fluor 350, Clone: 502430, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8370U100UG
This item is not returnable.
View return policy
Description
Ghrelin Receptor/GHSR Monoclonal specifically detects Ghrelin Receptor/GHSR in Human samples. It is validated for Flow Cytometry.Specifications
Ghrelin Receptor/GHSR | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_940799 | |
GHSR | |
NS0 mouse myeloma cell line transfected with human GHSR Accession # NP_940799 | |
RUO | |
2693 | |
Human | |
IgG1 |
Flow Cytometry | |
502430 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
GH-releasing peptide receptor, Ghrelin receptor, GHRP, GHS-R, growth hormone secretagogue receptor, growth hormone secretagogue receptor type 1 | |
Mouse | |
100 μg | |
Primary | |
Detects GHSR on transfectants by flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)