Learn More
Ghrelin Receptor/GHSR Mouse anti-Human, Alexa Fluor 405, Clone: 502430, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8370V100UG
Catalog No. FAB8370V100
Add to cart
This item is not returnable. View return policy
Description
Description
Ghrelin Receptor/GHSR Monoclonal antibody specifically detects Ghrelin Receptor/GHSR in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Ghrelin Receptor/GHSR
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
NP_940799
GHSR
NS0 mouse myeloma cell line transfected with human GHSR Accession # NP_940799
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects GHSR on transfectants by flow cytometry.
Flow Cytometry
502430
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
GH-releasing peptide receptor, Ghrelin receptor, GHRP, GHS-R, growth hormone secretagogue receptor, growth hormone secretagogue receptor type 1
Mouse
IgG1
RUO
Primary
2693
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)