Learn More
ZING™ Enterprises GHS Introduction Poster
ZING GHS Introduction Poster defines the Globally Harmonized System, the 3 major changes and how the changes benefit companies.
$46.35 - $116.40
Specifications
Material | Plastic |
---|---|
Quantity | 1 Ea. |
Mounting Type | Self-adhesive |
For Use With (Application) | Communication |
Height (English) | 24 in. |
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
18-002-994
|
ZING™ Enterprises
6036 |
Right to Understand |
Each for $46.35
|
|
|||||
18-002-995
|
ZING™ Enterprises
6037 |
Right to Understand - Labels |
Each for $46.35
|
|
|||||
18-002-996
|
ZING™ Enterprises
6038 |
Right to Understand - Safety Data Sheets |
Each for $46.35
|
|
|||||
18-002-997
|
ZING™ Enterprises
6070 |
Right to Understand ALL |
Pack of 3 for $116.40
|
|
|||||
Description
Poster is printed on recycled plastic to support sustainability initiatives and LEED credits, measure 18 in.H x 24 in.W. Made in the USA.
ZING GHS Labeling Poster defines the 6 labeling elements and 9 pictograms used in the Globally Harmonized System. Poster is printed on recycled plastic to support sustainability initiatives and LEED credits, measure 18 in.H x 24 in.W. Made in the USA.
ZING GHS Safety Data Sheet Poster defines the 16 section format used in the Globally Harmonized System. Poster is printed on recycled plastic to support sustainability initiatives and LEED credits, measure 18 in.H x 24 in.W. Made in the USA.
ZING GHS SDS Poster set supports company compliance and training programs related to the new GHS/Hazard Communication requirements. Set includes 3 posters (1) General Introduction Poster: defines the Globally Harmonized System, the 3 major changes and how the changes benefit companies, (2) GHS Labeling Poster: defines the 6 labeling elements and 9 pictograms used in the Globally Harmonized System, (3) GHS Safety Data Sheet Poster defines the 16 section format used in the Globally Harmonized System.
Specifications
Plastic | |
Self-adhesive | |
24 in. | |
English | |
Hazard Communication |
1 Ea. | |
Communication | |
18 in. | |
24 x 18 in. | |
Red on White |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.