Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ GHS Labeled Right-to-Know Safety-Vented Wash Bottles
Click to view available options
Capacity:
500 mL
1000 mL
Chemical Label:
ASSORTED
Acetone
Deionized Water
Dichloromethane
Ethanol
Ethyl Acetate
Isopropanol
Machine Oil
Methanol
Sodium Hypochlorite (Bleach)
Water
Closure Color:
Assorted
Blue
Green
Natural
Red
Yellow
Description
Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.
- Pre-printed bottles with chemical name and formula, GHS pictogram and signal word; National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
- Translucent LDPE bottles, Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
- Wide mouth 53mm (2 in.) for easy refilling
- Color-coded polypropylene closures aid in quick and easy identification
- Assortment bag includes one each: Acetone, Isopropanol, Bleach and Ethanol
Bottle with Label: TOLUENE
- Red LDPE bottle
- Does not include vented valve
Bottle with Label: DEIONIZED H2O, DISTILLED H20 and WATER
- Does not indicate hazardous chemical and does not have/require GHS pictogram or signal word
Bottle with Label: METHYL ETHYL KETONE
- Does not include vented valve
Compliance
Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA’s HazCom 2012 requirements – as pertains to Workplace Labels.
Specifications
Chemical Label | Deionized Water |
Closure Size | 53 mm |
Closure Type | Cap |
Closure Material | Polypropylene |
Through the Closure Dispensing | Yes |
Product Type | Safety Vented Wash Bottle |
Capacity | 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16.91 oz. |
Color | Natural |
Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction