Learn More
Fisherbrand™ GHS Labeled Right-to-Know Safety-Vented Wash Bottles
Unique valve prevents pressure build-up eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position
 View more versions of this product
Catalog No. 11-865-156
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
500mL
LDPE
Natural
MACHINE OIL
3/Bag
Wide mouth