Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Giα1 Mouse anti-Rat, Clone: 7H7, MilliporeSigma™

Mouse Monoclonal Antibody

Specifications

Antigen Giα1
Clone 7H7
Concentration 1.0 mg/mL
Applications Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
05555I100UG
View Documents
MilliporeSigma
05555I100UG
100 μg N/A
05555I25UG
View Documents
MilliporeSigma
05555I25UG
25 μg N/A
Description

Description

Giα1 Monoclonal antibody specifically detects Giα1 in Rat samples. It is validated for Western Blotting.
Specifications

Specifications

Giα1
1.0 mg/mL
Unconjugated
Mouse
Rat
KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal half of rat Gia-1.
Monoclonal
Protein G purified
7H7
Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG2bκ
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit