Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Giant Inflatable Nocturnal Creatures Products for Science Education

Introduce animal and insect behavior, characteristics and special adaptations of nocturnal creatures — the firefly even lights up!

Manufacturer:  Learning Resources, IncSupplier Diversity Partner LER1795

Catalog No. S02192


Add to cart

Alert:

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.


Includes:

  • Four creatures, Activity Guide, hanging hooks and repair kit


Description

Description

  • Great classroom decoration
  • Set includes firefly, bat, spider and moth
  • Firefly features built-in LED light powered by three button-cell 1.5V batteries (included)
  • Durable vinyl wipes clean
  • Largest creature (moth) measures 30L × 25 in.H (76 × 63.5cm)

Order Info

Grades K+

Specifications

Specifications

Largest creature (moth) measures 30L x 25 in.H (76 x 63.5 cm)
Giant Inflatable Nocturnal Creatures
Firefly (with built-in LED light powered by three button-cell 1.5V batteries), bat, spider and moth; Activity guide; Hanging hooks; Repair kit
Introduce animal and insect behavior, characteristics and special adaptations of nocturnal creatures
K
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit