Learn More
Giant-Size Models Products for Science Education
Manufacturer:  Molecular Model Enterprise SRM 19V513
 View more versions of this product
Catalog No. S44044A
Add to cart
Description
Description
Construct simple to complex organic and inorganic molecular structures with demonstration-size models.
  • Atom centers are finished in standard colors
  • Bond positions are clearly identified
  • Connecting rods are color coded for identifying bond types
  • Bond lengths are furnished in two scales: giant structures for viewing in the lecture hall and smaller structures for the average classroom
Includes:
  • Fifty 5cm diameter atom centers
  • 104 wood connecting rods
  • Variety of flexible bonds and orbital connectors
  • Convenient storage box with compartments
  • Well-illustrated instruction book
Optional Orbital Attachments. For building accurate orbital models in three dimensions.
  • Lightweight orbitals attach to the atom centers
  • Adaptable for teaching advanced concepts in orbital theory
  • Pi bonds fit over ends of orbitals to show overlap of p orbitals
Contain 32 large 10cm orbitals, 18 pi bonds, compartmentalized storage container, and illustrated instructions.
Specifications
Specifications
Fifty 5cm dia. atom centers, 104 Wood connecting rods, variety of flexible bonds and orbital connectors, convenient storage box with compartments, well-illustrated instruction book
Orbital attachment