Learn More
Andwin ScientificSupplier Diversity Partner GIANT SPINBAR 28.6X159MM DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner 58948978EA
The 28x159mm giant spinbar is designed for stirring large volumes up to 50 gallons. It features a polygon shape adding turbulence and molded center pivot ring minimizing contact between the bar and vessel, reducing friction and chattering.
Catalog No. 50-112-6070